Pierce Erase-It® Background Eliminator Kit, Thermo Scientific
Supplier: Thermo Scientific
21065
CAPI21065EA 424.89 CAD
CAPI21065
Pierce Erase-It® Background Eliminator Kit, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The background eliminator reduces background signal evenly over any film within minutes
  • Fast, easy background elimination from film
  • Eliminates unwanted background from overexposed, speckled or shaded films
  • No need for time-consuming re-exposures to find the optimal image
  • No need to re-optimise assay reagents in order to obtain the optimal image
The eliminator will work with any application that requires X-ray film exposures, and it is compatible with both newly developed films or films that have been stored for years. Simply immerse the exposed film in the working solution, watch for the desired image, and stop the reaction by rinsing the film with water. There is no need to re-expose the blot or even completely re-run the experiment.
High background, shading, overexposed bands and speckling are problems inherent to film exposure. The Erase-It® kit can correct these problems without the need to re-expose the blot to film or re-do the experiment. Erase-It® solution can be used with newly exposed films or exposed films that have been stored. It can also be used with any brand of film.