Pierce QuantaBlu™ Fluorogenic Peroxidase Substrates, Thermo Scientific
Supplier: Thermo Scientific
15169
CAPI15169EA 747.69 CAD
CAPI15169
Pierce QuantaBlu™ Fluorogenic Peroxidase Substrates, Thermo Scientific
Electrophoresis Reagents Western Blotting Reagents
The substrate generates a blue fluorescent product upon reaction with peroxidase and does not photobleach.
  • More sensitive than TMB, OPD or ABTS substrates
  • Large dynamic range (4 log peroxidase concentration range)
  • Working solution stable for 24 hours
  • Large stokes shift; excitation/emission maxima of 325/420; range of 315 to 345/370 to 460
  • Does not photobleach
QuantaBlu™ substrate also allows for stopped, nonstopped, and kinetic assays to be performed with low-level detection sensitivity and high signal-to-noise ratios.
Delivery information: Kit includes QuantaBlu™ substrate solution (250 mL), QuantaBlu™ peroxide solution (30 mL), QuantaBlu stop solution (275 mL).
Order Now
SPECIFICATIONS
Environmentally Preferable