Potassium iodide 99, granules, BAKER ANALYZED® ACS

Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Danger

Synonyms: Dipotassium diiodide, Kali iodide

BAKR3162-05 BAKR3162-07 BAKR3162-09
CAJT3162-1CS 3282.37 CAD
CAJT3162-1 CAJT3162-5 CAJT3162-07 CAJT3162-9 CAJT3162-11
Potassium iodide 99, granules, BAKER ANALYZED® ACS
Potassium iodide

Potassium Iodide in granular form.


ACS Reagent.

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.10 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Meets ACS Reagent Chemical Requirements
For Laboratory,Research,or Manufacturing Use
Assay (KI) ≥99.0 %
Insoluble Matter ≤0.005 %
Loss on Drying at 150°C ≤0.2 %
pH of 5% Solution at 25°C 6.0 - 9.2
Chloride and Bromide (as Cl) ≤0.01 %
Phosphate (PO₄) ≤0.001 %
Sulfate (SO₄) ≤0.005 %
Barium (Ba) ≤0.002 %
Calcium (Ca) ≤0.002 %
Magnesium (Mg) ≤0.001 %
Sodium (Na) ≤0.005 %
Iodate (IO₃) ≤3 ppm
Trace Impurities - ACS - Heavy Metals (as Pb) ≤5 ppm
Trace Impurities - Iron (Fe) ≤2.0 ppm
Meets Reagent Specifications for testing USP/NF monographs

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR