Potassium iodide 99%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: Dipotassium diiodide, Kali iodide

A12704-0B A12704-0I A12704-18 A12704-30
CAAAAA12704-0BEA 423.44 CAD
CAAAAA12704-0B CAAAAA12704-0I CAAAAA12704-18 CAAAAA12704-30
Potassium iodide 99%
Potassium iodide

Purity: 99% CAS No.: 7681-11-0 Molecular Formula: KI Formula Weight: 166.01 Refractive Index: 1.677 Odor: Odorless Boiling Point: 1330° Merck Reference: 14,7643 MDL No.: MFCD00011405

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.12
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 14,7643

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR