Anti-PDI Mouse Monoclonal Antibody

Supplier: Enzo Life Sciences
ADI-SPA-891-D ADI-SPA-891-F SPA-891J
89133-402EA 415.54 CAD
89133-402 89133-404 CA95046-300
Anti-PDI Mouse Monoclonal Antibody
Antibodies
The mammalian PDI (Protein disulfide-isomerase) family encompasses several highly divergent proteins which are involved in the processing and maturation of secretory proteins in the endoplasmic reticulum by catalyzing the rearrangement of disulfide bonds.

  • For Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Western Blot, EM

Recognizes human, mouse, rat, bovine, dog, chicken, guinea pig, hamster, monkey, pig, rabbit, sheep and Xenopus PDI. Detects a band of ~58kDa by Western blot.

Type: Primary
Antigen: PDI
Clonality: Monoclonal
Clone:
More Product Information
Order Now

SPECIFICATIONS

Antigen symbol PDI PDI
Environmentally Preferable
Antigen name Protein Disulphide-Isomerase Protein Disulphide-Isomerase
Antigen synonyms protein disulfide isomerase|Eps1p|PDI1|CaO19.12595|PDI1P protein disulfide isomerase|Eps1p|PDI1|CaO19.12595|PDI1P
Antibody type Primary Primary
Clonality Monoclonal Monoclonal
Reactivity Human Human
Host Mouse Mouse
Isotype IgG1 IgG1
Western blot Yes Yes
ImmunoChemistry Yes Yes
ImmunoFluorescence Yes Yes
Form Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Purification Protein G affinity purified

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR