Difco™ MYP Agar Base, BD
Supplier: BD CANADA MICROBIOLOGY CA
281010
CA90004-024EA 231.68 CAD
CA90004-024
Difco™ MYP Agar Base, BD
Media for Microbiology Prepared Media for Microbiology
MYP Agar Base with added supplements is used for isolation and identification of pathogenic Staphylococci and Bacillus species.
  • Peptic digest and meat extract provide nitrogen source
  • Phenol red indicates mannitol fermentation
  • Egg yolk emulsion helps differentiate lecithinase-producing colonies
MYP Agar Base is recommended by BIS committee under the specifications IS:5887-1976.