Difco™ Tellurite Enrichment Medium, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
277910
CA90003-984PK 805.27 CAD
CA90003-984
Difco™ Tellurite Enrichment Medium, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Used with Baird-Parker Agar Base for the isolation of S
aureus. The enrichment consists of 30% egg yolk suspension with 0.15% potassium tellurite.