Difco™ Starch Agar, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
272100
CA90003-890EA 413.36 CAD
CA90003-890
Difco™ Starch Agar, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Starch Agar is used for cultivating microorganisms being tested for starch hydrolysis.
  • Beef extract provides nitrogen, vitamins, carbon, and amino acids
  • Dehydrated appearance: Light beige, free-flowing
  • Agar is the solidifying agent
Starch Agar is used in differentiating microorganisms based on the starch hydrolysis test.