Difco™ KF Streptococcus Agar, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
249610
CA90003-436EA 369.99 CAD
CA90003-436
Difco™ KF Streptococcus Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Use with TTC or TTC Solution 1%, if desired For selective enumeration of fecal streptococci.