Difco™ SOB Medium, BD
Supplier: BD CANADA MICROBIOLOGY CA
244310
CA90003-336EA 459.89 CAD
CA90003-336
Difco™ SOB Medium, BD
Media for Microbiology Prepared Media for Microbiology
For cultivation of molecular genetic strains of E.coli.
  • Dehydrated culture media