Difco™ Azide Dextrose Broth, BD
Supplier: BD CANADA MICROBIOLOGY CA
238710
CA90003-102EA 387.05 CAD
CA90003-102
Difco™ Azide Dextrose Broth, BD
Media for Microbiology Dehydrated Media for Microbiology
Azide Dextrose Broth is used for cultivating streptococci in water and wastewater.
  • Beef extract and peptones provide essential nutrients
  • Dextrose is a fermentable carbohydrate
  • Sodium chloride maintains osmotic equilibrium
  • Sodium azide inhibits cytochrome oxidase in gram-negative bacteria
Azide Dextrose Broth is specified for use in the presumptive test of water and wastewater for fecal streptococci by the Multiple-Tube Technique.
Order Now
SPECIFICATIONS
Environmentally Preferable