BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
231673 231705 231541 231762 231251 231267 231292 231293 231301 231311 231313 231315 231324 231328 231342 231344 231352 231353 231539 231544 231601 231603 231605 231609 231619 231621 231645 231653 231666 231672 231674 231678 231692 231693 231694 231696 231703 231704 231706 232174 231709 231752 232175 231299 231314 231319 231321 231569
CA90002-494EA 38.96 CAD
CA90002-494 CA90002-532 CA-90002-328 CA-90002-506 CA-90002-544 CA-90002-582 CA90002-080 CA90002-226 CA90002-232 CA90002-242 CA90002-252 CA90002-254 CA90002-258 CA90002-264 CA90002-266 CA90002-270 CA90002-272 CA90002-274 CA90002-278 CA90002-286 CA90002-290 CA90002-292 CA90002-296 CA90002-298 CA90002-300 CA90002-308 CA90002-310 CA90002-326 CA90002-330 CA90002-390 CA90002-392 CA90002-394 CA90002-402 CA90002-404 CA90002-416 CA90002-420 CA90002-422 CA90002-440 CA90002-452 CA90002-468 CA90002-488 CA90002-492 CA90002-496 CA90002-500 CA90002-508 CA90002-514 CA90002-516 CA90002-518 CA90002-522 CA90002-528 CA90002-530 CA90002-534 CA90002-596 CA90010-200 CA95027-258 CA90002-598 CA90002-262L CA90002-276L CA90002-280L CA90002-284L CA90002-348L
BD BBL™ Sensi-Disc™ Antimicrobial Susceptibility Test Discs, BD Diagnostics
Microbiology Test Systems Microorganism Tests
The 6 mm diameter (¹/₄") discs are made from high quality absorbent paper that has been impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents. Discs are clearly marked on both sides with letters and numbers designating the agent and the drug content.
Ordering information: Discs are packaged 50 discs per cartridge. The last disc in each cartridge is marked with an 'X' and contains the agent as coded. Cartridges are for use in BD BBL™ Sensi-Disc™ dispensers.