Difco™ Nutrient Agar with MUG, BD
Supplier: BD CANADA MICROBIOLOGY CA
223200
CA90001-748EA 761.04 CAD
CA90001-748 CA90001-734
Difco™ Nutrient Agar with MUG, BD
Media for Microbiology Dehydrated Media for Microbiology
Nutrient Agar with MUG is used for detecting and enumerating Escherichia coli in water.
  • Beef extract provides carbohydrates, vitamins, nitrogen, and salts
  • Peptones provide a source of nitrogen
  • Agar is the solidifying agent
Rapid quantification and verification may be achieved with the membrane filtration procedure by transferring the membrane from a total-coliform or fecal-coliform positive sample to a Nutrient Agar substrate containing 4-methylumbelliferyl-β-D-glucuronide (MUG).