BBL™ Gram Basic Fuchsin, BD
Supplier: BD CANADA MICROBIOLOGY CA
212545
CA90000-716EA 1014.72 CAD
CA90000-716
BBL™ Gram Basic Fuchsin, BD
Media for Microbiology Dehydrated Media for Microbiology
Gram stain reagents are used to stain microorganisms from cultures or specimens by the differential Gram method.
  • Differential Gram method
  • Stains more intensely than safranin
Basic fuchsin stains many Gram-negative bacteria more intensely than safranin, making them easier to see.
Order Now
SPECIFICATIONS
Environmentally Preferable