BBL™ Enterococcosel™ Broth, BD Biosciences
Supplier: BD CANADA MICROBIOLOGY CA
212207
CA90000-530EA 796.3 CAD
CA90000-530
BBL™ Enterococcosel™ Broth, BD Biosciences
Media for Microbiology Dehydrated Media for Microbiology
Enterococcosel™ Broth is used for the presumptive identification of enterococci.
  • Selective medium
  • Effectively differentiates enterococci
The combination of esculin and a rather low concentration of bile in the presence of azide permits the selection of enterococci and differentiation by esculin hydrolysis.