Difco™ Malt Extract Broth, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
211320
CA90000-150EA 263.06 CAD
CA90000-150
Difco™ Malt Extract Broth, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malt Extract Broth is used for cultivating yeasts and molds.
  • Malt extract provides carbon, protein, and nutrient sources for essential growth
  • Maltose is an energy source
  • Dextrose is included as a source of fermentable carbohydrate
  • Yeast extract provides vitamins and additional nitrogen and carbon
The acidic pH of Malt Extract Broth allows for the optimal growth of molds and yeasts while restricting bacterial growth.