Difco™ Malt Extract Agar, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
211220
CA90000-120EA 348.47 CAD
CA90000-120
Difco™ Malt Extract Agar, BD Diagnostics
Media for Microbiology Prepared Media for Microbiology
Malt Extract Agar is used for isolating, cultivating, and enumerating yeasts and molds.
  • Maltose is an energy source
  • Dextrin and glycerol are sources of carbon
  • Peptone provides a nitrogen source
  • Agar is the solidifying agent
The acidic pH of Malt Extract Agar allows for the optimal growth of molds and yeasts while restricting bacterial growth.