Lugol's iodine solution concentrate for Gram staining
Supplier: AZER SCIENTIFIC CA
ES815
CA89130-198CS 2080.61 CAD
CA89130-198
Lugol's iodine solution concentrate for Gram staining
Lugol's iodine solution
Used as the mordant in the Gram stain protocol
Added to diluent when product is ready to use