Giemsa's stain Azure B

Supplier: AZER SCIENTIFIC CA

Synonyms: Azure mixture sicc. Giemsa stain, Azur-Eosin-Methylene blue acc. Giemsa, Giemsa's azur-eosin-methylene blue

ES5544
CA89130-176EA 507.32 CAD
CA89130-176
Giemsa's stain Azure B
Giemsa's stain

Stain for blood smears or bone marrow specimens.


  • Giemsa stain solution
  • For in-vitro diagnostic use


Giemsa stain is a classical blood film stain for peripheral blood smears and bone marrow specimens. It is used to visualize chromosomes, stain fungus histoplasma and identify mast cells.

MDL Number: MFCD00081642
CAS Number: 51811-82-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR