Buffer, EDTA Buffer Solution pH 8,5 (0,5 mol/L)
Supplier: G-Biosciences
R013
CA82023-102EA 161.6 CAD
CA82023-102
Buffer, EDTA Buffer Solution pH 8,5 (0,5 mol/L)
Buffers Cloning Buffers
A DNase and RNase free solution of 0.5M EDTA.