Caspase-3 Substrate VII, Fluorogenic, Calbiochem®

Supplier: MilliporeSigma

264151-1MG 264151-5MG
CA80051-372EA 358.76 CAD
CA80051-372 CA80051-374
Caspase-3 Substrate VII, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents

Fluorogenic caspase-3 substrate


Also acts as a substrate for granzyme-B processed ICE-LAP3, caspase-6, caspase-7, caspase-8, and caspase-10. Excitation maximum: ~400nm; emission maximum: ~505nm.

Order Now

SPECIFICATIONS

Environmentally Preferable
Form White lyophilized solid
Molecular weight 729.6
Purity 95% by HPLC
Solubility DMSO

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR