Caspase-6 Substrate III, Fluorogenic, Calbiochem®
Supplier: MilliporeSigma
218763-5MG
CA80050-850EA 919.64 CAD
CA80050-850
Caspase-6 Substrate III, Fluorogenic, Calbiochem®
Electrophoresis Reagents Western Blotting Reagents
A fluorogenic substrate for caspase-6 (Mch2)
Reaction can be monitored visually or quantitatively by a blue to green shift in fluorescence upon cleavage of the AFC fluorophore. Excitation maximum: ~400nm; emission maximum: ~505nm.