Anti-Ki-67 Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
RP026-01-RUO RP026-05-RUO RP026-RUO
CA76634-994EA 127.93 CAD
CA76634-994 CA76634-996 CA76634-998
Anti-Ki-67 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a nuclear antigen present in proliferating human cells. It can be used to evaluate the Ki-67 labeling index in various tumors.
Order Now

SPECIFICATIONS

Antigen name Ki-67
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR