Anti-Flk-1 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
PDR171-RUO
CA76634-672EA 567.39 CAD
CA76634-672
Anti-Flk-1 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 180 kDa protein, known as Flk-1, KDR or vascular endothelial growth factor receptor 2 (VEGFR2). Flk-1 is a cell membrane receptor kinase. Flk-1 is a high affinity receptor for vascular endothelial growth factor and is putatively involved in the growth of endothelial cells and angiogenesis.