Anti-Heliocobacter Pylori Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
PDR053-RUO
CA76634-652EA 415.4 CAD
CA76634-652
Anti-Heliocobacter Pylori Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with the whole H. pylori. The antibody stains the bacteria present on the surface of the epithelium or in the cytoplasm of the epithelial cells. H. pylori is known to cause peptic ulcer and chronic gastritis in humans.
Order Now

SPECIFICATIONS

Antigen name Heliocobacter pylori
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR