Anti-Granzyme B Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
PDR050-RUO
CA76634-650EA 382.28 CAD
CA76634-650
Anti-Granzyme B Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a 32 kDa protein. Granzymes are neutral serine proteases, which are stored in specialized lytic granules of cytotoxic T lymphocytes (CTL) and in natural killer (NK) cells. A number of granzymes (A to G) have been isolated and cloned. Human granzyme B is involved in target cell apoptosis during lymphocyte-mediated cytotoxicity. This antibody is useful in the localization of granzyme B containing lytic granules and for the characterization of activated CTL or NK cells.
Order Now

SPECIFICATIONS

Environmentally Preferable
Antigen name Granzyme B
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR