Anti-Ki-67 Rabbit Polyclonal Antibody
Supplier: Diagnostic Biosystems
PDR048-RUO
CA76634-646EA 374.76 CAD
CA76634-646
Anti-Ki-67 Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with a nuclear antigen present in proliferating human cells. It can be used to evaluate the Ki-67 labeling index in various tumors.