Anti-HCG Rabbit Polyclonal Antibody

Supplier: Diagnostic Biosystems
PDR030-RUO
CA76634-636EA 227.27 CAD
CA76634-636
Anti-HCG Rabbit Polyclonal Antibody
Antibodies
This antibody reacts with hCG. It stains strongly with the human placenta and placental trophoblasts.
Order Now

SPECIFICATIONS

Antigen name HCG
Environmentally Preferable
Antibody type Primary
Clonality Polyclonal
Conjugation Unconjugated
Reactivity Human
Host Rabbit
Isotype

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR