Anti-HHV8 Rat Monoclonal Antibody [clone: LN53]

Supplier: Diagnostic Biosystems
PDM395-RUO
CA76634-506EA 405.38 CAD
CA76634-506
Anti-HHV8 Rat Monoclonal Antibody [clone: LN53]
Antibodies
This antibody stains with ORF-73 of human herpesvirus 8 (HHV8). HHV8 is the likely etiological agent of Kaposi sarcoma (KS). HHV8 encodes a latent nuclear antigen (LNA), which is the product of the viral gene ORF 73. LNA is capable of forming a complex with retinoblastoma susceptibility gene product, which may be related to its oncogenic activity.
Order Now

SPECIFICATIONS

Antigen name HHV8
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone LN53
Reactivity Human
Host Rat
Isotype IgG2c

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR