Anti-BCL-2 Oncoprotein Mouse Monoclonal Antibody [clone: 124]

Supplier: Diagnostic Biosystems
PDM016-RUO
CA76634-280EA 466.7 CAD
CA76634-280
Anti-BCL-2 Oncoprotein Mouse Monoclonal Antibody [clone: 124]
Antibodies
This antibody recognizes a protein of 25 to 26kDa, identified as bcl-2 β-oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 alpha oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 alpha protein, whereas the normal or hyperplastic germinal centers are negative. This antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas thatMore Product Information
Order Now

SPECIFICATIONS

Antigen name BCL-2 oncoprotein
Environmentally Preferable
Antibody type Primary
Clonality Monoclonal
Conjugation Unconjugated
Clone 124
Reactivity Human
Host Mouse
Isotype IgG1 kappa

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR