Anti-CD45 LCA Mouse Monoclonal Antibody [clone: 2B11+PD7/26]
Supplier: Diagnostic Biosystems
PDM009-RUO
CA76634-268EA 369.63 CAD
CA76634-268
Anti-CD45 LCA Mouse Monoclonal Antibody [clone: 2B11+PD7/26]
Antibodies
CD45 is useful in differential diagnosis of lymphoid tumors from non-hematopoietic undifferentiated neoplasms.