Anti-HLA-DR β chain of DP DQ and DR Mouse Monoclonal Antibody [clone: CR3/43]
Supplier: Diagnostic Biosystems
MOB069-01-RUO
CA76633-218EA 144.79 CAD
CA76633-218
Anti-HLA-DR β chain of DP DQ and DR Mouse Monoclonal Antibody [clone: CR3/43]
Antibodies
This antibody reacts with the β chain of all products of the gene sub regions DP, DQ, and DR. It stains B cells, interdigitating reticulum cells, Langerhans cells and many macrophages. This antibody does not react with normal T cell and polymorphs, but stains activated T cells.