Latitude® Precast Agarose Gels, Lonza
Supplier: Lonza
57226 57234 57246 57230
CA12001-468EA 438.01 CAD
CA12001-468 CA12001-472 CA12001-474 CA12001-484 CA12002-152
Latitude® Precast Agarose Gels, Lonza
Electrophoresis Gels Precast Gels
Ideal for checking restriction digests and PCR products.
HT gels are designed for high throughput screening applications. They can be run on most large submerged electrophoresis systems and feature landscape gel orientations. Midigels feature portrait gel orientations. RNA midigels also feature portrait gel orientations, but are ideal for Northern blotting and RNA electrophoresis. Both the HT gels and the DNA midigels are pre-stained with 0.5 µg/ml ethidium bromide. The RNA midigels are not pre-stained, but are compatible with ethidium bromide, SYBR® Green II, and GelStar® nucleic acidMore Product Information