Tissue Marking Dyes, Azer Scientific
Supplier: AZER SCIENTIFIC CA
ESTMD-2BK ESTMD-8BK ESTMD-2B ESTMD-8B ESTMD-2GN ESTMD-8GN ESTMD-8O ESTMD-2R ESTMD-8R ESTMD-2V ESTMD-2Y
CA11008-414EA 180.63 CAD
CA11008-414 CA11008-416 CA11008-418 CA11008-420 CA11008-422 CA11008-424 CA11008-428 CA11008-430 CA11008-432 CA11008-434 CA11008-438
Tissue Marking Dyes, Azer Scientific
Stains Histology Stains
Mark critical margins and areas of interest on tissue sections with these assorted colors of tissue marking dye.
  • Available in black and six vibrant colors
  • Colors: black, blue, green, orange, red, violet, yellow
  • Saturated color quickly tranfers to tissue
  • Ready-to-use formula
Mark margins and areas of interest on tissue samples with this tissue marking dye.
Ordering information: Dyes are available individually in 2 oz. and 8 oz. bottles.
Delivery information: Kits include 2 oz. bottles of each color dye, a base and applicator sticks.