Mounting Medium, Azer Scientific
Supplier: AZER SCIENTIFIC CA
ES723 ES724
CA10821-472CS 2587.8 CAD
CA10821-472 CA10821-474 CA10821-476 CA10821-478
Mounting Medium, Azer Scientific
Stains Microscopy and staining reagents
Adhesive mounting medium with permanent clarity for long term slide storage.
  • Traditional formula or quick drying acrylic
  • Low viscosity for easy cover glass mounting
Non-aqueous S-mounting medium formula is compatible with xylene and most xylene substitutes, dries quickly and and will not crack or yellow over time.
Traditional mounting medium is soluble in xylene and toluene, dries optically clear and will resist photobleaching and stain fading.