T4 DNA Ligase Reaction Buffer, New England Biolabs
Supplier: New England Biolabs (NEB)
B0202S
CA101227-368EA 42 CAD
CA101227-368
T4 DNA Ligase Reaction Buffer, New England Biolabs
Buffers Cloning Buffers
New England Biolabs supplies a 10X reaction buffer with all of its enzymes
At a 1X concentration this reaction buffer assures optimal activity of the enzyme.
Order Now
SPECIFICATIONS
Environmentally Preferable