BD Taxo™ X Factor Discs, BD Diagnostic Systems
Supplier: BD CANADA MICROBIOLOGY CA
231729
CA-90002-554EA 39.83 CAD
CA-90002-554
BD Taxo™ X Factor Discs, BD Diagnostic Systems
Microbiology Test Systems Microorganism Tests
BD Taxo™ X Factor Discs are used for differentiating Haemophilus species.
These paper discs are round, brown 1/4” with “x” printed on both sides.