Tryptic Soy Broth (TSB) Dehydrated and Prepared Culture Media, BD Diagnostics
Supplier: BD CANADA MICROBIOLOGY CA
221823 211768 211771 211822 211823 211825 221815 221351 221716 211772
CA-90001-542PK 706.71 CAD
CA-90001-542 CA90000-344 CA90000-346 CA90000-372 CA90000-374 CA90000-378 CA90001-224 CA90001-534 CA-90001-332 CA-90001-496 CA90000-376 CA90001-494 CA90000-348
Tryptic Soy Broth (TSB) Dehydrated and Prepared Culture Media, BD Diagnostics
Media for Microbiology Dehydrated Media for Microbiology
Tryptic soy broth is used in microbiology laboratories as a culture broth to grow aerobic bacteria.
  • Used in a wide variety of procedures
  • May be used for suspension and enrichment
  • For cultivation of strains isolated on other media
With the addition of 6.5% sodium chloride, differentiation between salt-tolerant and salt-intolerant species is achieved.
Order Now
SPECIFICATIONS
Environmentally Preferable