BBL™ Urea Agar, BD Life Sciences
Supplier: BD CANADA MICROBIOLOGY CA
221097
CA-90001-230PK 639.43 CAD
CA-90001-230 CA90001-228
BBL™ Urea Agar, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Urea Agar is a differential medium for members of the Enterobacteriaceae on the basis of their ability to produce urease.
  • Peptone and dextrose provide nutrients
  • Reduced buffer content helps promote rapid growth
  • Utilized urea produces ammonia
When organisms utilize urea, ammonia is formed during incubation, which makes the reaction of these media alkaline, producing a pink-red color. Consequently, urease production may be detected by the change in the phenol red indicator.