N(α)-Cbz-L-histidine

Supplier: BACHEM AMERICAS INC

Synonyms: Cbz-His-OH, N(α)-Benzyloxycarbonyl-L-histidine, N-Cbz-L-histidine, Z-His-OH

4000625.0005 4000625.0025
C-1945.0005BAEA 175.66 CAD
C-1945.0005BA C-1945.0025BA
N(α)-Cbz-L-histidine
N(α)-Cbz-L-histidine

Sequence: Z-His-OH


The atoms of the imidazole ring of histidine can be numbered in two different ways. Bachem as well as organic chemists designate the position of N-τ as 1 and the position of N-π as 3, whereas biochemists generally number N-π, the nitrogen atom adjacent to the side chain, as 1, and N-τ as 3.

Formula: C₁₄H₁₅N₃O₄
MW: 289.3 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00065960
CAS Number: 14997-58-1
EINECS: 239-084-0

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR