Potassium iodide ACS, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Dipotassium diiodide, Kali iodide

BDH9264-2.5KG BDH9264-125G BDH9264-500G
BDH9264-2.5KGCS 6171.02 CAD
BDH9264-2.5KG BDH9264-125G BDH9264-500G
Potassium iodide ACS, VWR Chemicals BDH®
Potassium iodide

Meets reagent specifications for testing USP/NF monographs.


Colorless or white solid

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Density: 3.13 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Barium <= 0.002 %
Calcium <= 0.002 %
Chloride & Bromide <= 0.01 %
Heavy Metals (as Pb) <= 0.0005 %
Insolubles <= 0.005 %
Iodate <= 0.0003 %
Iron <= 0.0003 %
Loss on Drying <= 0.2 %
Magnesium <= 0.001 %
pH (5%, Water) @25C 6.0 - 9.2
Phosphate <= 0.001 %
Purity >= 99.0 %
Sodium <= 0.005 %
Sulfate <= 0.005 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR