Potassium iodide 5% w/v aqueous solution Stabilised zA, VWR Chemicals BDH®

Supplier: VWR International

Synonyms: Dipotassium diiodide, Kali iodide

BDH7458-1
BDH7458-1CS 2669.24 CAD
BDH7458-1
Potassium iodide 5% w/v aqueous solution Stabilised zA, VWR Chemicals BDH®
Potassium iodide

Made with potassium iodide, potassium hydroxide solution and deionized water.

Formula: Kl
MW: 166 g/mol
Boiling Pt: 1330
Melting Pt: 686
Storage Temperature: Ambient
MDL Number: MFCD00011405
CAS Number: 7681-11-0
EINECS: 231-659-4
Merck Index: 13,07727

Order Now

Specification Test Results

Appearance/color Clear liquid
Shelf-life 1 year


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR