Fmoc-Lys(Ac)-OH

Supplier: BACHEM AMERICAS INC

4034189.0001 4034189.0005 4034189.0025
B-3520.0001BAEA 197.47 CAD
B-3520.0001BA B-3520.0005BA B-3520.0025BA
Fmoc-Lys(Ac)-OH
Fmoc-Lys(Ac)-OH

Sequence: Fmoc-Lys(Ac)-OH


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₃H₂₆N₂O₅
Storage Temperature: Freezer
MDL Number: MFCD00077409
CAS Number: 159766-56-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR