Fmoc-D-Asp(OPp)-OH

Supplier: BACHEM AMERICAS INC

4027583.0001 4027583.0005
B-2955.0001BAEA 800.3 CAD
B-2955.0001BA B-2955.0005BA
Fmoc-D-Asp(OPp)-OH
Fmoc-D-Asp(OPp)-OH

Sequence: Fmoc-D-Asp(2-phenylisopropyl ester)-OH


Asp-NH₂ is also called isoasparagine, so our Asp-NH₂ derivatives are assembled in the "Isoasparagine" subfamily.


Synonym(s): Fmoc-D-Asp(OPp)-OH

Formula: C₂₈H₂₇NO₆
Storage Temperature: Freezer
MDL Number: MFCD00798646
CAS Number: 214852-39-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR