Fmoc-D-Arg(Boc)2-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-D-Arg(Boc)₂-OH

4027707.0001
B-2940.0001BAEA 1114.07 CAD
B-2940.0001BA
Fmoc-D-Arg(Boc)2-OH
Fmoc-D-Arg(Boc)2-OH

Sequence: Fmoc-D-Arg(Boc)₂-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.

Formula: C₃₁H₄₀N₄O₈
MW: 596.681 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00798643
CAS Number: 214852-34-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR