Fmoc-Hyp-OH

Supplier: BACHEM AMERICAS INC

4025531.0005 4025531.0025
B-2800.0005BAEA 232.35 CAD
B-2800.0005BA B-2800.0025BA
Fmoc-Hyp-OH
Fmoc-Hyp-OH

Sequence: Fmoc-Hyp-OH


Hyp denotes L-trans-hydroxyproline.

Formula: C₂₀H₁₉NO₅
Storage Temperature: Freezer
MDL Number: MFCD00151929
CAS Number: 88050-17-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR