Fmoc-L-3,3-diphenylalanine

Supplier: BACHEM AMERICAS INC

4030246.0250 4030246.1000 4030246.5000
B-2725.0250BAEA 232.35 CAD
B-2725.0250BA B-2725.1000BA B-2725.5000BA
Fmoc-L-3,3-diphenylalanine
Fmoc-L-3,3-diphenylalanine

Sequence: Fmoc-β,β-diphenyl-Ala-OH


Besides Ala derivatives, Bachem offers a number of derivatives of β-substituted alanines, amongst them: t-Butylalanine Cyanoalanine Cyclohexylalanine (Cha) Cyclopropylalanine 2-Quinolylalanine β-Halogenated Alanines
Dip derivatives.


Synonym(s): Fmoc-Dip-OH#Fmoc-β-phenyl-Phe-OH#Fmoc-3,3-diphenyl-L-alanine

Formula: C₃₀H₂₅NO₄
Storage Temperature: Ambient
MDL Number: MFCD00672337
CAS Number: 201484-50-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR