Fmoc-Lys(Me3)-OH chloride

Supplier: BACHEM AMERICAS INC

Synonyms: Fmoc-Lys(Me)₃-OH chloride, Fmoc-Lys(Me)3-OH Chloride, Fmoc-Lys(Me)3-OH Chloride

4026139.0100 4026139.0250 4026139.1000
B-2685.0100BAEA 413.45 CAD
B-2685.0100BA B-2685.0250BA B-2685.1000BA
Fmoc-Lys(Me3)-OH chloride
Fmoc-Lys(Me3)-OH chloride

Sequence: Fmoc-Lys(Me)₃-OH chloride


For biotinylated lysine derivatives or chromophores and fluorophores coupled to Lys please see the subfamily "Nε-labeled Lysine".

Formula: C₂₄H₃₁ClN₂O₄
MW: 446.972 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00672333
CAS Number: 201004-29-7

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR