Fmoc-Arg(Pbf)-OH

Supplier: BACHEM AMERICAS INC

Synonyms: Nα-Fmoc-Nω-Pbf-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulfonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine, Nα-Fmoc-Nomega-(2,2,4,6,7-pentamethyl-2,3-dihydrobenzo[b]furan-5-ylsulphonyl)-L-arginine, Nɑ-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nω-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulphonyl)-L-arginine

4089151.0005 4089151.0025 4089151.0100
B-2375.0005BAEA 233.1 CAD
B-2375.0005BA B-2375.0025BA B-2375.0100BA
Fmoc-Arg(Pbf)-OH
Nalpha-[(9H-Fluoren-9-ylmethoxy)carbonyl]-Nomega-(2,2,4,6,7-pentamethyldihydrobenzofuran-5-sulfonyl)-L-arginine

Sequence: Fmoc-Arg(Pbf)-OH


Bachem additionally offers decarboxy-Arg: Agmatine · sulfate Q-1880.


Pbf protection is especially recommended for multiple-Arg- and Trp-containinng peptides. TFA cleavage is best performed in the presence of a silane scavenger.

Formula: C₃₄H₄₀N₄O₇S
MW: 648.78 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00235804
CAS Number: 154445-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR