Fmoc-Gln(Mtt)-OH

Supplier: BACHEM AMERICAS INC

4017008.0001 4017008.0005 4017008.0025
B-2050.0001BAEA 175.66 CAD
B-2050.0001BA B-2050.0005BA B-2050.0025BA
Fmoc-Gln(Mtt)-OH
Fmoc-Gln(Mtt)-OH

Sequence: Fmoc-Gln(Mtt)-OH


For isoglutamine derivatives please see the family "Glutamic Acid".

Formula: C₄₀H₃₆N₂O₅
Storage Temperature: Refrigerator
MDL Number: MFCD00237163
CAS Number: 144317-23-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR